Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAS2R9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567753
Description
Predicted to react with Mouse and Rat samples.
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13. Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells.
Specifications
TAS2R9 | |
Polyclonal | |
Unconjugated | |
TAS2R9 | |
T2R9; TAS2R9; taste 2 receptor member 9; taste receptor family B member 6; Taste receptor type 2 member 9; taste receptor, family B, member 6; taste receptor, type 2, member 9; TRB6 | |
Rabbit | |
affinity chromatography | |
RUO | |
50835 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NYW1 | |
TAS2R9 | |
Synthesized peptide derived from human TAS2R9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction