Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TASK-5/KCNK15 Antibody, Novus Biologicals™
SDP

Catalog No. NB396553 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396553 25 μL
NBP239038 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396553 Supplier Novus Biologicals Supplier No. NBP23903825UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TASK-5/KCNK15 Polyclonal specifically detects TASK-5/KCNK15 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TASK-5/KCNK15
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9H427
Gene Alias Acid-sensitive potassium channel protein TASK-5, dJ781B1.1, K2p15.1, KCNK11, KCNK14, KIAA0237, KT3.3, potassium channel subfamily K member 15, potassium channel, subfamily K, member 14, potassium channel, subfamily K, member 15, TASK-5, TASK5Two pore potassium channel KT3.3, TWIK-related acid-sensitive K(+) channel 5, Two pore K(+) channel KT3.3
Gene Symbols KCNK15
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GALRRKFGFSAEDYRELERLALQAE
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60598
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.