Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TASP 0390325
GREENER_CHOICESDP

Catalog No. 572950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-295-0 50 mg
57-291-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-295-0 Supplier Tocris Bioscience™ Supplier No. 5729/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective V1B receptor antagonist; orally bioavailable

TASP 0390325 is a potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.

Specifications

CAS 1642187-96-9
Quantity 50 mg
Target Vasopressin Receptor
Molecular Formula C25H29ClFN5O4.HCl
Formula Weight 554.44g/mol
Product Type Antagonist

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.