Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TASP 0433864
GREENER_CHOICESDP

Catalog No. 536210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-621-0 10 mg
53-625-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-621-0 Supplier Tocris Bioscience™ Supplier No. 5362/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective mGlu2 receptor positive allosteric modulator

TASP 0433864 is a selective mGlu2 receptor positive allosteric modulator (EC50 values are 199 and 206 nM for human and rat receptors, respectively). Exhibits negligible activity against a range of neurotransmitter targets, except 5-HT2B and MAO-B (72 and 92% inhibition at 10 μM, respectively). Potentiates inhibitory effect of DCG IV (Cat. No. 0975) on excitatory postsynaptic potentials in hippocampal slices and ketamine- and methamphetamine-induced increases in locomotor activity in rats and mice.

Specifications

Chemical Name or Material (2S)-2-[[4-(1,1-Dimethylethyl)phenoxy]methyl]-2,3-dihydro-5-methylimidazo[2,1-b]oxazole-6-carboxamide
CAS 1431980-60-7
Synonym mGlu2 receptor positive allosteric modulator
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C18H23N3O3
Formula Weight 329.39
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.