Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TASP1 Monoclonal Antibody (OTI1H6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502257S


Description
TASP1 Monoclonal antibody specifically detects TASP1 in Human samples. It is validated for Western BlotSpecifications
TASP1 | |
Monoclonal | |
0.54 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O43347 | |
Mouse | |
Affinity chromatography | |
RUO | |
4440 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI1H6 | |
Unconjugated | |
MSI1 | |
4930485D02Rik; AW986064; C20orf13; dJ585I14.2; FLJ20212; MGC39159; RGD1308591; si:dkey-38k3.3; Tasp1; taspase 1; taspase, threonine aspartase 1; taspase, threonine aspartase, 1; taspase-1; Threonine aspartase 1; Threonine aspartase 1-like protein; Threonine aspartase subunit alpha; Threonine aspartase subunit beta | |
Full length human recombinant protein of human MSI1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction