Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TASP1 Antibody (OTI1H6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274444H
Description
TASP1 Monoclonal specifically detects TASP1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TASP1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
TASP1 | |
Full length recombinant protein of human TASP1 (NP_060184) produced in HEK293T cells. | |
0.1 mL | |
55617.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1H6 | |
Western Blot | |
C20orf13taspase 1, chromosome 20 open reading frame 13, dJ585I14.2, EC 3.4.25.-, FLJ20212, MGC39159, taspase, threonine aspartase, 1, Taspase-1, threonine aspartase 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction