Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544679 Supplier Invitrogen™ Supplier No. PA544679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QNKQTLLVEF LWSHTTESMC VGYMSAQDGK AKTHISRLPP GAVAGQSVAI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintenance of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TASP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TASP1
Gene Accession No. Q9H6P5
Gene Alias 4930485D02Rik; AW986064; C20orf13; dJ585I14.2; FLJ20212; MGC39159; RGD1308591; si:dkey-38k3.3; Tasp1; taspase 1; taspase, threonine aspartase 1; taspase, threonine aspartase, 1; taspase-1; Threonine aspartase 1; Threonine aspartase 1-like protein; Threonine aspartase subunit alpha; Threonine aspartase subunit beta
Gene Symbols TASP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TASP1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55617
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.