Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TAT |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17928620
![]() |
Novus Biologicals
NBP17928620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179286
![]() |
Novus Biologicals
NBP179286 |
100 μL |
Each for $487.50
|
|
|||||
Description
TAT Polyclonal specifically detects TAT in Rat samples. It is validated for Western Blot.Specifications
TAT | |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.6.1.5, L-tyrosine:2-oxoglutarate aminotransferase, tyrosine aminotransferase, tyrosine aminotransferase, cytosolic | |
TAT | |
IgG |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Rabbit | |
NP_036800 | |
6898 | |
The immunogen for this antibody is Tat. Peptide sequence PGLQPVRPSGAMYLMVGIEMEHFPEFENDVEFTERLIAEQAVHCLPATCF. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title