Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAT complex Monoclonal Antibody (C1), Invitrogen™

Catalog No. FEMA551122
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551122 20 μL
FEMA551121 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551122 Supplier Invitrogen™ Supplier No. MA551122
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Thrombin-antithrombin complex (TAT complex) is a protein complex formed by the binding of thrombin, a serine protease involved in blood clotting, and antithrombin, a serine protease inhibitor that regulates thrombin activity. This complex plays a crucial role in maintaining the balance between blood clotting and preventing excessive clotting, which can lead to thrombosis. The complex is formed when antithrombin binds to thrombin, inhibiting its activity and preventing the formation of blood clots.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAT complex
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene SERPINC1
Gene Accession No. P00734, P01008
Gene Alias TAT
Gene Symbols F2, SERPINC1
Host Species Mouse
Immunogen Recombinant Human TAT protein, Ile364-Glu622 + GGGGS*3 + His33-Lys464 (Accession #P00734 & P01008).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2147, 462
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.