Learn More
Invitrogen™ TAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113872
Description
Immunogen sequence: PGLQPVRPSG AMYLMVGIEM EHFPEFENDV EFTERLIAEQ AVHCLPATCF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.
TAT is a mitochondrial protein tyrosine aminotransferase which is present in the liver and catalyzes the conversion of L-tyrosine into p-hydroxyphenylpyruvate.
Specifications
| TAT | |
| Polyclonal | |
| Unconjugated | |
| TAT | |
| 2.6.1.5; L-tyrosine:2-oxoglutarate aminotransferase; Tat; testis tissue sperm-binding protein Li 34a; tyrosine aminotransferase; tyrosine aminotransferase, cytosolic; tyrosine transaminase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24813 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P04694 | |
| TAT | |
| Immunogen sequence: PGLQPVRPSG AMYLMVGIEM EHFPEFENDV EFTERLIAEQ AVHCLPATCF (aa 361-410). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.