Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAT Polyclonal Antibody, Huabio

Catalog No. 502544631
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4631 Supplier HUABIO RESEARCH INC Supplier No. HA500016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAT Polyclonal antibody specifically detects TAT in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen TAT
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TAT
Gene Accession No. P04694, P17735, Q8QZR1
Gene Alias 2.6.1.5; L-tyrosine:2-oxoglutarate aminotransferase; Tat; testis tissue sperm-binding protein Li 34a; tyrosine aminotransferase; tyrosine aminotransferase, cytosolic; tyrosine transaminase
Host Species Rabbit
Immunogen Recombinant protein within human TAT aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234724, 24813, 6898
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.