Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tat-SF1 Recombinant Rabbit Monoclonal Antibody (JE54-89)
GREENER_CHOICE

Catalog No. PIMA536200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536200 Supplier Invitrogen™ Supplier No. MA536200
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Tat-SF1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tat-SF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE54-89
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Htatsf1
Gene Accession No. O43719, Q8BGC0
Gene Alias 1600023H17Rik; 2600017A12Rik; 2700077B20Rik; cofactor required for Tat activation of HIV-1 transcription; dJ196E23.2; HIV TAT specific factor 1; HIV Tat-specific factor 1; HIV Tat-specific factor 1 homolog; HIV-1 Tat specific factor 1; Htatsf1; RGD1563377; RP1-196E23.2; TATSF1; Tat-SF1
Gene Symbols Htatsf1
Host Species Rabbit
Immunogen Recombinant protein within Human Tat-SF1 aa 1-140.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27336, 72459
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.