Learn More
TATA binding protein TBP Antibody (C.15200002), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259208
Catalog No. NBP259208
Only null left
Add to Cart
Description
Description
TATA binding protein TBP Monoclonal specifically detects TATA binding protein TBP in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Chromatin Immunoprecipitation Sequencing, Knockdown Validated.
Specifications
Specifications
TATA binding protein TBP
Monoclonal
Unconjugated
PBS with 0.05% Sodium Azide
TBP
The exact immunogen is propietary information.
100 μg
Transcription Factors and Regulators
6908
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, ChIP Assay, KnockDown
C.15200002
Western Blot 1:500, Chromatin Immunoprecipitation (ChIP) 4-5 ug/IP, Chromatin Immunoprecipitation Sequencing 4 - 5 ug/IP, Knockdown Validated
GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit
Mouse
ammonium sulphate purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only