Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TATA binding protein TBP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238610
Description
TATA binding protein TBP Polyclonal specifically detects TATA binding protein TBP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TATA binding protein TBP | |
Polyclonal | |
Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
P20226 | |
TBP | |
This antibody was developed against a recombinant protein corresponding to amino acids: QQAVAAAAVQQSTSQQATQGTSGQAPQLFHSQTLTTAPLPGTTPLYPS | |
0.1 mL | |
Transcription Factors and Regulators | |
6908 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction