Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TATA binding protein TBP/TBP Monoclonal Antibody (6G8), Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33227M

This item is not returnable.
View return policy
Description
TATA binding protein TBP/TBP Monoclonal antibody specifically detects TATA binding protein TBP/TBP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TATA binding protein TBP/TBP | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P20226, P29037 | |
Mouse | |
Protein G | |
RUO | |
117526, 21374, 6908 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
6G8 | |
Unconjugated | |
Tbp | |
Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100) | |
Recombinant human TBP protein | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction