Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TATA Element Modulatory Factor 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP190117
Description
TATA Element Modulatory Factor 1 Polyclonal specifically detects TATA Element Modulatory Factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
TATA Element Modulatory Factor 1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
TATA element modulatory factor 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
7110 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TMF1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TSTTSDIEVLDHESVISESSASSRQETTDSKSSLHLMQTSFQLLSASACPEYNRLDDFQKLTESCCSSDAFERIDSFSVQSLDSRSVSEINSDDELSGKGYALVPIIVNSSTPKSKTVESAEGKSEEVNETLVIPTEEAEMEESG | |
0.1 mL | |
Primary | |
Specificity of human TATA Element Modulatory Factor 1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction