Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TATDN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554853
Description
Immunogen sequence: CGEFEKNNPD LYLKELLNLA ENNKGKVVAI GECGLDFDRL QFCPKDTQLK YFEKQFELSE QTKLP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
TATDN1 is a protein coding gene. Putative deoxyribonuclease.
Specifications
TATDN1 | |
Polyclonal | |
Unconjugated | |
TATDN1 | |
2310079P03Rik; CDA11; hepatocarcinoma high expression protein; LOC690068; Putative deoxyribonuclease TATDN1; similar to TatD DNase domain containing 1; TatD DNase domain containing 1; Tatdn1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
690068, 69694, 83940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P1N9, Q6P8M1 | |
TATDN1 | |
Recombinant protein corresponding to Human TATDN1. Recombinant protein control fragment (Product #RP-93819). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction