Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TATDN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584140
Description
Immunogen sequence: MSRFKFIDIG INLTDPMFRG IYRGVQKHQD DLQDVIGRAV EIGVKKFMIT GGNLQDSKDA LHLAQTNGMF.
TATDN1 is a protein coding gene. Putative deoxyribonuclease.
Specifications
TATDN1 | |
Polyclonal | |
Unconjugated | |
TATDN1 | |
2310079P03Rik; CDA11; hepatocarcinoma high expression protein; LOC690068; Putative deoxyribonuclease TATDN1; similar to TatD DNase domain containing 1; TatD DNase domain containing 1; Tatdn1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P1N9 | |
TATDN1 | |
Recombinant protein corresponding to Human TATDN1. Recombinant protein control fragment (Product #RP-104959). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction