Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TATDN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588351 Supplier Invitrogen™ Supplier No. PA588351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRFKFIDIG INLTDPMFRG IYRGVQKHQD DLQDVIGRAV EIGVKKFMIT GGNLQDSKDA LHLAQTNGMF FSTVGCHPTR CGEFEKNNPD LYLKELLNLA ENNKGKVVAI GECGLDFDRL QFCPKDTQLK YFEKQFELSE QTKLPMFLHC RNSHAEFLDI MKRNRDRCVG GVVHSFDGTK EAAAALIDLD LYIGFNGCSL KTEANLEVLK SIPSEKLMIE TDAPWCGVKS THAGSKYIRT AFPTKKKWES GHCLKDRNEP CHIIQILEIM SAVRDEDPLE LANTLYNNTI KVFFPGI; Positive Samples: LO2, A-549, A-431; Cellular Location: Nucleus.

TATDN1 is a protein coding gene. Putative deoxyribonuclease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TATDN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TATDN1
Gene Accession No. Q6P1N9
Gene Alias 2310079P03Rik; CDA11; hepatocarcinoma high expression protein; LOC690068; Putative deoxyribonuclease TATDN1; similar to TatD DNase domain containing 1; TatD DNase domain containing 1; Tatdn1
Gene Symbols TATDN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-297 of human TATDN1 (NP_1144151).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83940
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.