Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TATDN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528490
Description
Recommended positive controls: NT2D1, SK-N-SH, mouse testis. Predicted reactivity: Mouse (86%), Rat (87%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
TATDN3 | |
Polyclonal | |
Unconjugated | |
TATDN3 | |
1500010M24Rik; Putative deoxyribonuclease TATDN3; TatD DNase domain containing 3; TATDN3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
128387, 68972 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q17R31, Q3U1C6 | |
TATDN3 | |
Recombinant protein fragment corresponding to a region within amino acids 56 and 242 of Human TATDN3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction