Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tau Antibody (1D5) - Pre-formed Fibrils, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279825
Description
Tau Monoclonal specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
Tau | |
Monoclonal | |
Unconjugated | |
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
MAPT | |
Human Recombinant Tau441 (2N4R), P301S mutant Protein Pre-formed Fibrils | |
0.1 mg | |
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
Detects Multiple Bands. Antibody detects monomer under denaturing conditions but preferentially detects fibril under native conditions (dot blot). | |
Store at -20°C. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot | |
1D5 | |
Western Blot 1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Dot Blot 1:100-1:2000 | |
DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction