Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Mouse anti-Mouse, Human, Unconjugated, Clone: 2A1-2E1, MilliporeSigma™

Catalog No. MABN2472100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2472-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2472-100 Supplier MilliporeSigma Supplier No. MABN2472100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tau, clone 2A1-2E1, Cat. No. MABN2472, is a highly specific mouse monoclonal antibody that targets Microtubule-associated protein tau and has been tested for use in Immunohistochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2A1-2E1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Microtubule-associated protein tau;Neurofibrillary tangle protein;Paired helical filament-tau;PHF-tau
Gene Symbols MAPT;MAPTL;MTBT1;TAU
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal half of human Microtubule-associated protein tau.
Molecular Weight of Antigen ∽70 kDa observed; 78.93 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_058519
Test Specificity Clone 2A1-2E1 detects Microtubule-associated protein tau in human and murine brain. It targets an epitope within 19 amino acids from the N-termnal half.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.