Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Mouse anti-Rat, Mouse, Human, Clone: TNT-2, MilliporeSigma™

Catalog No. MABN2406MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2406MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2406MI Supplier MilliporeSigma Supplier No. MABN2406
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tau Antibody, clone TNT-2 is a highly specific mouse monoclonal antibody that targets Tau and has been tested in ELISA, Immunofluorescence, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Immunofluorescence, ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone TNT-2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols MAPT,MAPTL,MTBT1,TAU
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human Tau.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_058519
Test Specificity Clone TNT-2 specirfically detects tau in human, mouse, and rat brain. It targets a sequence of 17 amino acids from the N-terminal region that is common to all isoforms (1-6) of tau.
Target Species Rat, Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.