Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tau Oligomeric Complex Antibody (TOC1), Janelia Fluor™ 646 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320163JF646
Description
Tau Oligomeric Complex Monoclonal antibody specifically detects Tau Oligomeric Complex in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
Tau Oligomeric Complex | |
Monoclonal | |
Janelia Fluor 646 | |
Tau Oligomeric Complex 1, TOC1, TOC-1 | |
The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
TOC1 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction