Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tau, oligomeric, Mouse, Unlabeled, Clone: TOMA-1,

Catalog No. MABN819 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN819 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN819 Supplier MilliporeSigma™ Supplier No. MABN819
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Microtubule-associated protein tau (UniProt P10636; also known as Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau) is encoded by the MAPT (also known as TAU, MAPT1, MTBTL) gene (Gene ID 4137) in human. In Alzheimer's disease (AD) pathology, accumulation of the microtubule-associated protein tau takes place primarily in the neurons. Tau accumulates in both the somatodendritic and axonal domains of neurons. Tau also accumulates in the soma as neurofibrillary tangles (NFTs). Cell death and synaptic lesions occur independently of NFT formation, and research indicates that NFT formation alone is insufficient for neurodegeneration, suggesting that soluble tau aggregates may be the more toxic and pathologically significant tau species. Tau oligomers are neurotoxic when applied extracellularly to cultured neuronal cells, and tau oligomers (but not fibrils) induce neurodegeneration and synaptic and mitochondrial dysfunction in vivo. Moreove, researchers are able to use tau oligomers as a reliable biomarker to differentiate AD brains from age-matched non-AD brains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau, oligomeric
Applications ELISA, Immunofluorescence, Immunohistochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone TOMA-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2aκ in buffer containing PBS without preservatives.
Gene MAPT, TAU, MTBT1, MAPTL
Gene Accession No. P10636
Host Species Mouse
Immunogen Recombinant human Tau-441 (Tau-F, Tau-4, 2N4R isoform) oligomers (Lasagna-Reeves, C. A., et al. (2012). FASEB J. 26(5):1946-1959).
Purification Method Protein A Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_058519.3
Test Specificity Clone TOMA-1 (a.k.a. clone H12C10) is among more than 13 hybridomas derived from mice immunized with in vitro generated recombinant Tau-441 (Tau-F, Tau-4, 2N4R isoform) oligomers. These TOMAs are reported to specifically detect oligomeric tau without any significant reactivity toward monomeric tau, tau fibrils, Aβ oligomers, Aβ fibrils, or α-synuclein oligomers (Castillo-Carranza, D.L., et al. (2014). J. Neurosci. 34(12):4260-4272).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.