Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau (TauC4, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABN217825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN217825UL 25 μL
ABN2178100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN217825UL Supplier MilliporeSigma Supplier No. ABN217825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Tau (TauC4, 354-369), Cat. No. ABN2178, is a highly specific rabbit polyclonal antibody that targets TauC4 region of Tau and has been tested for use in Electron Microscopy and Western Blotting.

Microtubule-associated protein tau (UniProt: P10636; also known as Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau) is encoded by the MAPT (also known as MAPTL, MTBT1, TAU) gene (Gene ID: 4137) in human. Microtubule-associated protein tau is shown to be expressed in neurons. It is mostly found in axons, in the cytosol, and in association with plasma membrane components. Nine isoforms of Tau have been described that are generated by alternative splicing. Tau proteins promote microtubule assembly and stability and may also be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, thus serving as a linker protein between both. The short isoforms are reported to allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization. In Alzheimer disease (AD), the neuronal cytoskeleton in the brain is progressively disrupted and replaced by tangles of paired helical filaments (PHF) and straight filaments, mainly composed of hyperphosphorylated forms of Tau. In AD brain Tau accumulates in both the somatodendritic and axonal domains of neurons and it also accumulates in the soma as neurofibrillary tangles (NFTs). Tau oligomers are reported to be neurotoxic when applied extracellularly to cultured neuronal cells and can induce neurodegeneration and synaptic and mitochondrial dysfunction in vivo. It has been reported that most of the C-terminus of Tau (aa 243-406) in PHFs can be cleaved by trypsin, however the TauC4 region is highly resistant to trypsin action. (Ref.: Taniguchi Watanabe, S., et al. (2016). Acta Neuropathol. 131(2); 267-280; Ando K., et al. (2010). Biochem. Soc. Trans. 38(4); 1001-1005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau (TauC4
Applications Immunomicroscopy, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Tau (TauC4, 354-369) in Western Blotting applications (Fitzpatrick, A.W.P., et. al. (2017) Nature. 547(7662):185-190; Taniguchi-Watanabe, S., et. al. (2016). Acta Neuropathol. 131(2):267-280).Electron Microscopy Analysis: A representative lot detected Tau (TauC4, 354-369) in Immunogold negative-stain Electron Microscopy applications (Fitzpatrick, A.W.P., et. al. (2017) Nature. 547(7662):185-190).
Formulation Rabbit polyclonal antiserum with 0.1% sodium azide.
Gene Alias Microtubule-associated protein tau;Neurofibrillary tangle protein;Paired helical filament-tau;PHF-tau
Gene Symbols MAPT;MAPTL;MTBT1;TAU
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal half of human Microtubule-associated protein tau, isoform 8 (Tau-F).
Molecular Weight of Antigen ∽55 kDa observed; 45.85 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005901
Test Specificity This rabbit polyclonal antibody specifically detects TauC4 region in human Tau protein.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.