Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine, Tocris Bioscience™
SDP

Catalog No. 02091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-091-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-091-G Supplier R&D Systems Supplier No. 0209/1G

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-selective, endogenous glycine receptor partial agonist

One of the most abundant free amino acids in the brain. A partial agonist at the inhibitory glycine receptor. May be used in protocols for the generation of retinal pigment epithelial cells from hPSCs

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
PubChem CID 1123
ChEBI CHEBI:15891
IUPAC Name 2-aminoethane-1-sulfonic acid
SMILES NCCS(O)(=O)=O

Specifications

Quantity 1 g
Formula Weight 125.14
Percent Purity >98%
Chemical Name or Material Taurine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.