Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurocholic Acid Sodium Salt, MP Biomedicals™

Catalog No. mp210293090
Click to view available options
Quantity:
500 g

Used for protein solubilization. Taurocholic Acid Sodium Salt, MP Biomedicals is an anionic detergent that is the conjugation product of cholic acid with taurine. It is the principal constituent of the bile of carnivorous animals.

Biochemical actions:

  • Non-sulfated bile salt
  • Physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp)
  • Na(+)/taurocholate cotransporter (NTCP)
  • MRP3 into the hepatic carnalicular
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-42-6
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.688
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
Synonym sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
PubChem CID 131632374
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Physical Form Powder
Melting Point 230°C (decomposition)(literature)
pH 6 at 7.5 (1% aq. soln.)
Quantity 500 g
Solubility Information Soluble in water (100mg/mL).
Formula Weight 537.7
Percent Purity ≥97%
Grade Practical
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 3-11mM
Recommended Storage Store at Room Temperature (15°C to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.