Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tauroursodeoxycholic Acid, Sodium Salt, Calbiochem™,

Catalog No. p-7082988 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 14605-22-2
Molecular Formula C26H45NO6S
Molecular Weight (g/mol) 499.71
MDL Number MFCD00065451
InChI Key BHTRKEVKTKCXOH-AYSJQVDDSA-N
Synonym Sodium Tauroursodeoxycholate
PubChem CID 9848818
ChEBI CHEBI:80774
IUPAC Name 2-[(4R)-4-[(1R,4S,5aS,7R,9aS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]ethane-1-sulfonic acid
SMILES C[C@H](CCC(=O)NCCS(O)(=O)=O)[C@H]1CCC2C3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)C3CC[C@]12C

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥97% (TLC)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.