Learn More
Invitrogen™ TAX1BP1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total TAX1BP1 protein.
Specifications
Specifications
| Antigen | TAX1BP1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | TAX1BP1 |
| Gene Accession No. | Q86VP1 |
| Gene Alias | 1200003J11Rik; 1700069J21Rik; Aa1076; AA930106; CALCOCO3; D6Ertd404e; D6Ertd772e; liver regeneration-related protein 2; liver regeneration-related protein LRRG004; PRO0105; T6BP; Tax1 (human T cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 binding protein 1; tax1-binding protein 1; Tax1-binding protein 1 homolog; TAX1BP1; TRAF6-binding protein; TRAF6-binding protein homolog; TXBP151 |
| Gene Symbols | TAX1BP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.