Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAX1BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555076 Supplier Invitrogen™ Supplier No. PA555076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADAVAELKLN AMKKDQDKTD TLEHELRREV EDLKLRLQMA ADHYKEKFKE CQRLQKQINK LSDQSANNNN VFTKKTGNQQ KVNDASVNTD PATSASTVDV KPSPSAAEAD FDIVTKGQVC EMTKEIADKT EKYNK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

This gene encodes a HTLV-1 tax1 binding protein. The encoded protein interacts with TNFAIP3, and inhibits TNF-induced apoptosis by mediating the TNFAIP3 anti-apoptotic activity. Degradation of this protein by caspase-3-like family proteins is associated with apoptosis induced by TNF. This protein may also have a role in the inhibition of inflammatory signaling pathways. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAX1BP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAX1BP1
Gene Accession No. Q86VP1
Gene Alias 1200003J11Rik; 1700069J21Rik; Aa1076; AA930106; CALCOCO3; D6Ertd404e; D6Ertd772e; liver regeneration-related protein 2; liver regeneration-related protein LRRG004; PRO0105; T6BP; Tax1 (human T cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 binding protein 1; tax1-binding protein 1; Tax1-binding protein 1 homolog; TAX1BP1; TRAF6-binding protein; TRAF6-binding protein homolog; TXBP151
Gene Symbols TAX1BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAX1BP1. Recombinant protein control fragment (Product #RP-101865).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.