Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAX1BP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566068
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6068 Supplier Bethyl Laboratories, Inc Supplier No. A303791A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

A TRAF-interacting protein that specifically associates with TRAF6, T6BP shares an overall 50% identity and 59% similarity to a recently identified chicken protein MDP62, which has been implicated in promoting neurite outgrowth. T6BP-TRAF6 complex formation is dependent on the presence of the IL-1 receptor-associated kinase (IRAK).

Specifications

Antigen TAX1BP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TAX1BP1
Gene Accession No. Q86VP1
Gene Alias CALCOCO3, T6BP, TXBP151
Gene Symbols TAX1BP1
Host Species Rabbit
Immunogen Between 550 and 600
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8887
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.