Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAX1BP1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | TAX1BP1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
TAX1BP1 Polyclonal specifically detects TAX1BP1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TAX1BP1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Rat | |
T6BP, Tax1 (human T-cell leukemia virus type I) binding protein 1, tax1-binding protein 1, TRAF6-binding protein, TXBP151CALCOCO3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 709-789 of human TAX1BP1 (NP_006015.4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
8887 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title