Learn More
TAX1BP3 Monoclonal Antibody (1H5)

Description
TAX1BP3 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | TAX1BP3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1H5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TAX1BP3 |
| Gene Accession No. | O14907 |
| Gene Alias | 1300011C24Rik; C20orf100; EMBL:AAH97976.1}; GCX1; glutaminase-interacting protein 3; Tax interaction protein 1; Tax1 (human T cell leukemia virus type I) binding protein 3; Tax1 (human T-cell leukemia virus type I) binding protein 3; Tax1 binding protein 3; tax1-binding protein 3; TAX1BP3; tax1bp3 {ECO:0000312; tax-interacting protein 1; TIP1; TIP-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.