Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAX1BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564090 Supplier Invitrogen™ Supplier No. PA564090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAEIAGLQIG DKIMQVNGWD MTMVTHDQAR KRLTKRSEEV VRLLVTRQSL QKAVQQSMLS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

May regulate a number of protein-protein interactions by competing for PDZ domain binding sites. Binds CTNNB1 and may thereby act as an inhibitor of the Wnt signaling pathway. Competes with LIN7A for KCNJ4 binding, and thereby promotes KCNJ4 internalization. May play a role in the Rho signaling pathway. May play a role in activation of CDC42 by the viral protein HPV16 E6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAX1BP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAX1BP3
Gene Accession No. O14907
Gene Alias 1300011C24Rik; C20orf100; EMBL:AAH97976.1}; GCX1; glutaminase-interacting protein 3; Tax interaction protein 1; Tax1 (human T cell leukemia virus type I) binding protein 3; Tax1 (human T-cell leukemia virus type I) binding protein 3; Tax1 binding protein 3; tax1-binding protein 3; TAX1BP3; tax1bp3 {ECO:0000312; tax-interacting protein 1; TIP1; TIP-1
Gene Symbols TAX1BP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAX1BP3. Recombinant protein control fragment (Product #RP-103490).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.