Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Taxifolin, ≥85% (HPLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
25 mg
100 mg
Description
Food and Beverages

Specifications
Specifications
Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
CAS | 480-18-2 |
Physical Form | Neat |
Linear Formula | C15H12O7 |
MDL Number | MFCD00006845 |
Synonym | (2 R,3 R)-3,3 ′,4 ′,5,7-Pentahydroxyflavanone; (2 R,3 R)-Dihydroquercetin |
Type | Analytical Standard |
Quantity | 100 mg |
Formula Weight | 304.25 |
Percent Purity | ≥85% (HPLC) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction