Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taxifolin, ≥85% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187708
Click to view available options
Quantity:
25 mg
100 mg

Taxifolin is a plant flavonoid, which may find applications in the treatment of coronary heart disease, angina pectoris, cerebral thrombosis, cerebral infarction and sequelae.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 480-18-2
Physical Form Neat
Linear Formula C15H12O7
MDL Number MFCD00006845
Synonym (2 R,3 R)-3,3 ′,4 ′,5,7-Pentahydroxyflavanone; (2 R,3 R)-Dihydroquercetin
Type Analytical Standard
Quantity 25 mg
Formula Weight 304.25
Percent Purity ≥85% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.