Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

taxilin gamma Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB266770 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB266770 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB266770 Supplier Novus Biologicals Supplier No. NBP222331H
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

taxilin gamma Polyclonal antibody specifically detects taxilin gamma in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen taxilin gamma
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias ELRG, factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription, FIAT, lipopolysaccharide-specific response protein 5, taxilin gamma
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 478 to 528 of human Taxilin Gamma using the numbering given in entry NP_060830.2 (GeneID 55787).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55787
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.