Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Taxol Janelia Fluor 526
GREENER_CHOICESDP

Catalog No. 7315100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7315100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7315100U Supplier Tocris Bioscience™ Supplier No. 7315/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Green fluorescent taxol derivative; probe for microtubule staining

Taxol Janelia Fluor™ 526 is a fluorogenic, green fluorescent taxol derivative for direct imaging of the microtubule cytoskeleton. Taxol Janelia Fluor™ 526 is fluorogenic: it fluoresces only once bound to microtubules, enabling hassle-free no-wash experiments. Excitation maximum = 531 nm; emission maximum = 548 nm.

Specifications

Chemical Name or Material 4-((8-(((1 S,2 R)-3-(((2 aR,4 S,4 AS,6 R,9 S,11 S,12 S,12 aR,12 bS)-12 b-Acetoxy-12-(benzoyloxy)-4,6,11-trihydroxy-4 A,8,13,13-tetramethyl-5-oxo-2 A,3,4,4 A,5,6,9,10,11,12,12 A,12 b-dodecahydro-1 H-7,11-methanocyclodeca[3,4 ]benzo[1,2-b]oxet-9-yl)oxy)-2-hydroxy-3-oxo-1-phen
CAS 2410613-93-1
Quantity 100 μg
Target Microtubule Probes
Molecular Formula C73H74F6N4O17.CF3CO2H
Purity 0.85
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.