Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Taxol Janelia Fluor 646
GREENER_CHOICE

Catalog No. 62661
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-661 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 62-661 Supplier Tocris Bioscience™ Supplier No. 6266/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Red fluorescent taxol derivative; probe for microtubule staining; protocol available

Taxol Janelia Fluor® 646 is a red fluorescent taxol derivative for direct imaging of the microtubule cytoskeleton. Taxol Janelia Fluor® 646 is fluorogenic: fluoresces only once bound to microtubules, enabling hassle-free no-wash experiments. Excitation maximum = 655 nm; emission maximum = 671 nm.

For more information on how Taxol-Janelia Fluor™ 646 may be used, see our protocol .

Specifications

Chemical Name or Material 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-(((2 S)-1-(((2 aR,4 S,4 AS,6 R,9 S,11 S,12 S,12 bS)-6,12 b-diacetoxy-9-(((2 R,3 S)-3-benzamido-2-hydroxy-3-phenylpropanoyl)oxy)-12-(benzoyloxy)-11-hydroxy-4 A,8,13,13-
Quantity 1 mg
Target Microtubule Probes
Molecular Formula C79H82N4O18Si
Formula Weight 1403.62
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.