Learn More
TAZ Mouse anti-Human, Clone: OTI5A4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810800

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy, hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction. Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known.Specifications
TAZ | |
Monoclonal | |
Unconjugated | |
TAZ | |
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6901 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5A4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16635 | |
TAZ | |
Human recombit protein fragment corresponding to amino acids 45-248 of human TAZ produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.