Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAZ / WWTR1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993284
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3284 10 μL
50-199-3283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3284 Supplier Active Motif Supplier No. 61266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy, hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction. Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known.

Specifications

Antigen TAZ / WWTR1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene TAZ
Gene Accession No. Q16635
Gene Alias 5031411C02Rik; 9130012G04Rik; AW107266; AW552613; Barth syndrome); BTHS; cardiomyopathy, dilated 3A (X-linked); CMD3A; EFE; EFE2; endocardial fibroelastosis 2; FLJ27390; G4.5; HGNC:11577; LVNCX; OTTHUMP00000061673; Protein G4.5; Tafazzin; tafazzin (cardiomyopathy, dilated 3A (X-linked); tafazzin (cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Barth syndrome); TAZ; Taz1; XAP-2; XX-FW83563B9.3
Gene Symbols TAZ
Host Species Rabbit
Immunogen TAZ / WWTR1 antibody was raised against a peptide corresponding to the C-terminal region of human TAZ / WWTR1.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6901
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.