Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBATA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570872
Description
This target displays homology in the following species: Human: 100%.
This gene encodes a protein that regulates thymic epithelial cell proliferation and thymus size. It has been identified as a ligand for the class I human leukocyte antigen (HLA-I) in thymus. Studies of the orthologous mouse protein suggest that it may also play a role in spermatid differentiation, as well as in neuronal morphogenesis and synaptic plasticity. Polymorphisms in this gene are associated with susceptibility for multiple sclerosis (MS). Alternative splicing results in multiple transcript variants.
Specifications
TBATA | |
Polyclonal | |
Unconjugated | |
TBATA | |
1700021K02Rik; AI428928; C10orf27; protein SPATIAL; Protein TBATA; RGD1561665; SPATIAL; stromal protein associated with thymii and lymph node; Stromal protein associated with thymii and lymph node homolog; stromal protein associated with thymii and lymph nodes; TBATA; thymus and testis specific nuclear protein; thymus, brain and testes associated; thymus, brain and testes-associated protein; Titest | |
Rabbit | |
Affinity Chromatography | |
RUO | |
219793 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96M53 | |
TBATA | |
synthetic peptide directed towards the C-terminal region of human TBATA. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction