Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105736 Supplier Invitrogen™ Supplier No. PA5105736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TBC1D1.

The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBC1D1
Gene Accession No. Q60949, Q86TI0
Gene Alias 1110062G02Rik; AI385682; AW555803; Kiaa1108; LYN; Lyncein; mKIAA1108; TBC; Tbc1; TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1; TBC1 domain family member 1; TBC1 domain family, member 1; Tbc1d1; tre-2/USP6, BUB2, cdc16) domain family, member 1
Gene Symbols TBC1D1
Host Species Rabbit
Immunogen A synthesized peptide derived from mouse Tbc1d1(Accession Q60949), corresponding to amino acid residues V611-N631.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23216, 57915
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.