Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520389
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520389 Supplier Invitrogen™ Supplier No. PA520389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Daudi cell lysate. PA5-20389 can be used with blocking peptide PEP-0506.

The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TBC1D1
Gene Accession No. Q86TI0
Gene Alias 1110062G02Rik; AI385682; AW555803; Kiaa1108; LYN; Lyncein; mKIAA1108; TBC; Tbc1; TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1; TBC1 domain family member 1; TBC1 domain family, member 1; Tbc1d1; tre-2/USP6, BUB2, cdc16) domain family, member 1
Gene Symbols TBC1D1
Host Species Rabbit
Immunogen a 22 amino acid peptide from near the carboxy terminus of human TBC1D1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23216
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.