Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559743
Description
Immunogen sequence: ENDLLNKRLK LDYEEITPCL KEVTTVWEKM LSTPGRSKIK FDMEKMHSAV GQGVPRHHRG EIWKFLAEQF HLKHQFPSKQ QPKD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.
Specifications
TBC1D1 | |
Polyclonal | |
Unconjugated | |
TBC1D1 | |
1110062G02Rik; AI385682; AW555803; Kiaa1108; LYN; Lyncein; mKIAA1108; TBC; Tbc1; TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1; TBC1 domain family member 1; TBC1 domain family, member 1; Tbc1d1; tre-2/USP6, BUB2, cdc16) domain family, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23216 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TI0 | |
TBC1D1 | |
Recombinant protein corresponding to Human TBC1D1. Recombinant protein control fragment (Product #RP-96601). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction