Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBC1D10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582312 Supplier Invitrogen™ Supplier No. PA582312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NWDKWMAKKH KKIRLRCQKG IPPSLRGRAW QYLSGGKVKL QQNPGKFDEL DMSPGDPKWL DVIERDLHRQ FPFHEMFVSR GGHGQQDLFR VLKAYTLYRP EEGYCQAQAP IAAVLLMHMP AEQAFWCLVQ ICEKYLPGYY SEKLEAIQLD G Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

Acts as GTPase-activating protein for RAB27A, but not for RAB2A, RAB3A, nor RAB4A. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D10A
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBC1D10A
Gene Accession No. Q9BXI6
Gene Alias AC004997.C22.2; AI447804; dJ130H16.1; dJ130H16.2; EBP50-PDX interactor of 64 kDa; EBP50-PDZ interactor of 64 kD; EPI64; EPI64 protein; mFLJ00288; Rab27A-GAP-alpha; TBC1 domain family member 10A; TBC1 domain family, member 10A; TBC1D10; Tbc1d10a
Gene Symbols TBC1D10A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBC1D10A. Recombinant protein control fragment (Product #RP-91346).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.