Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D10A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572110
Description
TBC1D10A acts as GTPase-activating protein for RAB27A, but not for RAB2A, RAB3A, nor RAB4A.
Specifications
TBC1D10A | |
Polyclonal | |
Unconjugated | |
TBC1D10A | |
TBC1 domain family member 10A; EBP50-PDX interactor of 64 kDa; EPI64 protein; Rab27A-GAP-alpha; TBC1D10A; EPI64; TBC1D10 | |
Rabbit | |
Protein A | |
RUO | |
103724, 83874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.42 mg/mL | |
PBS with 0.09% sodium azide | |
P58802, Q9BXI6 | |
TBC1D10A | |
KLH conjugated synthetic peptide between 239-267 amino acids from the Central region of human TBC1D10A | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction