Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D10B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548889
Description
Acts as GTPase-activating protein for RAB3A, RAB22A, RAB27A, AND RAB35. Does not act on RAB2A and RAB6A.
Specifications
| TBC1D10B | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q4KMP7 | |
| TBC1D10B | |
| KLH conjugated synthetic peptide between 781-808 amino acids from the C-terminal region of human TBC1D10B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TBC1D10B | |
| TBC1 domain family member 10B, Rab27A-GAP-beta, TBC1D10B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26000 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction