Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558479
Description
Immunogen sequence: MVGPEDAGAC SGRNPKLLPV PAPDPVGQDR KVIRATGGFG GGVGAVEPPE EADEEE Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.

Specifications
TBC1D12 | |
Polyclonal | |
Unconjugated | |
TBC1D12 | |
KIAA0608; TBC1 domain family member 12; TBC1 domain family, member 12; TBC1D12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23232 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60347 | |
TBC1D12 | |
Recombinant protein corresponding to Human TBC1D12. Recombinant protein control fragment (Product #RP-97060). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction